- Antigen name:Ly-6C
- Clonality:Monoclonal
- Clone:HK1.4
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
- Host:Rat
- Isotype:IgG2c kappa
- Reactivity:Mouse
- Gene ID:17067
- Pk:100 µG
Rat monoclonal antibody against mouse Ly-6C, PerCP-Cy5.5-conjugated.
- Endothelial cells
- Granulocytes
- Subsets
- Lymphocytes
- Macrophages
- Monocytes
This monoclonal antibody (clone HK1.4) reacts with mouse lymphocyte antigen 6 family member C (Ly-6C), a 14 - 17 kDa glycosylphosphatidylinositol (GPI)-linked protein of the Ly6 family. Ly-6C is expressed on monocytes, macrophages, endothelial cells, granulocytes, and a subset of lymphocytes. Use this recombinant monoclonal antibody in applications requiring detection of Ly-6C, such as flow cytometry (FC). It has been reported that Ly-6C is an indicator of memory CD8+ T cells. Ly-6C is regulated by interferon gamma and may play a role in the development and maturation of lymphocytes. Some variation of Ly-6C expression on CD4 and CD8 lymphocytes is found in different mouse strains. Mouse strains with the Ly-6.2 alloantigen have subsets of CD4 and CD8 cells that express Ly-6.2, whereas strains with the Ly-6.1 alloantigen possess only Ly-6.1+ CD8 subsets.
Type:
Antigen: Ly-6C
Clonality: Monoclonal
Clone: HK1.4
Conjugation: PerCP-Cy5.5
Epitope:
Host: Rat
Isotype: IgG2c kappa
Reactivity: Mouse