Order Entry
Denmark
ContactUsLinkComponent
Biochrom Biowave 3+ Colour Touch
Biochrom Biowave 3+ Colour Touch
Catalog # WAPR80-3007-57
Supplier:  Biochrom
undefined
Biochrom Biowave 3+ Colour Touch
Catalog # WAPR80-3007-57
Supplier:  Biochrom

Specifications

  • Light source:
    Xenon Lamp
  • Description:
    Biowave 3+ Colour Touch UV-visible spectrophotometer
  • W×D×H:
    260×390×100 mm
  • Wavelength range:
    190 - 1100 nm
  • Interfaces:
    Full colour icon driven touch screen
  • Spectral bandwidth:
    3 nm
  • Weight:
    3 kg
  • Pk:
    1

Specifications

About this item

The Biowave 3 series offers all the benefits and capabilities of a research grade Life Science spectrophotometer with robust design and reasonable cost that also makes it perfect for students.

  • The Biowave 3+ Color Touch is truly an all-encompassing life sciences spectrophotometer
  • From the most basic cell density or single wavelength reading, to low volume molecular quantification, this is the most versatile single beam UV/Visible instrument on the market
  • This compact and robust instrument is perfect for most Life Science laboratories, from undergraduate teaching to biotech and R&D, the Biowave 3+ CT can cover the full spectrum of needs

Pre-programmed methodologies for nucleic acid quantification (DNA, RNA and oligonucleotides), protein assays (BCA, Biuret, Bradford and Lowry) and for cell culture density (OD 600) measurements. The visualisation of the nucleic acid scan is particularly useful, especially for RNA samples where impurities may be present in the 230 nm region, yet not have an adverse effect on the A260/A280 ratio. The system is compatible with low volume UV cuvettes. For those samples that need the most precise results, the Biowave 3+ CT also comes loaded with a menu of commonly available fluorescent dyes to specifically target nucleic acids and proteins.

In kinetics mode, the basic plot of absorbance against time may be supplemented with the result for A/min, plus the correlation coefficient is also calculated for the duration of the assay. This slope may be multiplied automatically by a factor to convert it directly to rate of reaction. The 127 mm color touch screen allows users to distinguish multiple samples by color on graph overlays and the zoom and track functions gives the ability to focus on areas of interest.

Results may be downloaded to a USB flash drive in CSV file format or printed to an optional integrated graphical printer for a permanent record. Alternatively, results can be exported via a cable connection to a suitable PC running Print Via Computer (PVC) software or wireless accessory (optional). Comprehensive IQ/OQ documentation and service support is available for our Biowave instruments allowing the qualification of wavelength accuracy, wavelength repeatability, photometric accuracy, stray light, resolution, noise, and drift.