Order Entry
Denmark
ContactUsLinkComponent
Human Recombinant GM-CSF (E. coli-expressed)
Human Recombinant GM-CSF (E. coli-expressed)
Certificates
undefined
Human Recombinant GM-CSF (E. coli-expressed)

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein/peptide name:
    GM-CSF
  • Purity:
    ≥95%

Specifications

Frequently Bought Together