Order Entry
Denmark
ContactUsLinkComponent
Human Recombinant GDNF, ACF
Human Recombinant GDNF, ACF
Certificates
undefined
Human Recombinant GDNF, ACF

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    GDNF, ACF
  • Purity:
    ≥95%

Specifications

Frequently Bought Together