Order Entry
Denmark
ContactUsLinkComponent
Mouse Recombinant VEGF-164
Mouse Recombinant VEGF-164
Certificates
undefined
Mouse Recombinant VEGF-164

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Protein/peptide name:
    VEGF-164
  • Purity:
    ≥97 %

Specifications

Frequently Bought Together