Order Entry
Denmark
ContactUsLinkComponent
 
Human Recombinant Oncostatin M
 
undefined
Human Recombinant Oncostatin M

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    Oncostatin M
  • Purity:
    ≥95%

 

Frequently Bought Together