Order Entry
Denmark
ContactUsLinkComponent
 
Mouse Recombinant IL-2
 
undefined
Mouse Recombinant IL-2

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Protein/peptide name:
    IL-2
  • Purity:
    ≥95%

 

Frequently Bought Together