Human Recombinant Noggin (CHO-expressed)
Noggin binds to and antagonizes bone morphogenetic protein (BMP) ligands of the transforming growth factor beta (TGF-β) family. Noggin supports maintenance of undifferentiated human embryonic stem cells in vitro, and can be used to prevent spontaneous differentiation in the short term (Chaturvedi et al.). Noggin is essential for development of structures derived from ectoderm embryonic somite, skeletal patterning, and neurogenesis in vivo. It also influences chondrogenesis, osteogenesis, and joint formation (Krause et al.), and promotes dopaminergic differentiation of embryonic stem cells and subsequent survival of dopamine neurons (Chiba et al.).
: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Protein synonyms:SYM1|SYNS1|NOG
- Protein/peptide name:Noggin
- Purity:≥95%
Frequently Bought Together