Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Mouse Recombinant G-CSF

Marketsource
Mouse Recombinant G-CSF
Marketsource
 
Mouse Recombinant G-CSF
Mouse Recombinant G-CSF

 

 

Granulocyte colony-stimulating factor (G-CSF) is a member of the CSF family of glycoproteins that regulate hematopoietic cell proliferation, differentiation, and function. It is a key cytokine involved in the production of neutrophils and the stimulation of granulocyte colony formation from hematopoietic progenitor cells (Metcalf and Nicola). G-CSF causes a range of effects including a transient reduction of SDF-1 expression (Petit et al.), the activation of metalloproteases that cleave VCAM-1 (Levesque et al.), and the release of norepinephrine from the sympathetic nervous system (Katayama et al.), leading to the release or mobilization of hematopoietic stem cells from the bone marrow into the periphery. The G-CSF receptor is expressed on a variety of hematopoietic cells, including myeloid-committed progenitor cells, neutrophils, granulocytes, and monocytes. In addition to hematopoietic cells, G-CSF is also expressed in cardiomyocytes, neuronal cells, mesothelial cells, and endothelial cells. Mouse G-CSF was first purified from cultures of the WEHI-3B myelomonocytic leukemia cell line as the inducer of the terminal differentiation of WEHI-3B and other myeloid leukemia cell lines (Nicola et al.). It was later cloned in monkey COS cells from a cDNA library prepared with mRNA derived from mouse fibrosarcoma NFSA cells that produce G-CSF constitutively (Tsuchiya et al.). Binding of G-CSF to its receptor leads to activation of the JAK/STAT, MAPK, PI3K, and AKT signal transduction pathways.

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

 

 

 

  • Catalog No:
  • STMC78014.1
  • STMC78014
  • STMC78014.2
  • Protein/peptide name:
  • G-CSF
  • G-CSF
  • G-CSF
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Species:
  • Mouse
  • Mouse
  • Mouse
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%

 

Product Family Options

 

  • Pk
     
     
  • 10 µG
       
       
    1 * 1 Each
    1 * 1 Each - 1.652,00 kr
    1.652,00 kr
    1 * 1 Each
       1.652,00 kr

     
    Marketsource
     
    Specifications
    Cat. No.
    STMC78014.1
    Protein/peptide name
    G-CSF
    Protein/peptide type
    Recombinant
    Species
    Mouse
    Purity
    ≥95%
    Marketsource
  • 100 µG
       
       
    1 * 1 Each
    1 * 1 Each - 6.329,00 kr
    6.329,00 kr
    1 * 1 Each
       6.329,00 kr

     
    Marketsource
     
    Specifications
    Cat. No.
    STMC78014
    Protein/peptide name
    G-CSF
    Protein/peptide type
    Recombinant
    Species
    Mouse
    Purity
    ≥95%
    Marketsource
  • 1.000 µG
       
       
    1 * 1 Each
    1 * 1 Each - 30.611,00 kr
    30.611,00 kr
    1 * 1 Each
       30.611,00 kr

     
    Marketsource
     
    Specifications
    Cat. No.
    STMC78014.2
    Protein/peptide name
    G-CSF
    Protein/peptide type
    Recombinant
    Species
    Mouse
    Purity
    ≥95%
    Marketsource