Order Entry
Denmark
ContactUsLinkComponent
 
Mouse Recombinant MIG (CXCL9)
 
undefined
Mouse Recombinant MIG (CXCL9)

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Protein/peptide name:
    MIG (CXCL9)
  • Purity:
    ≥95%

 

Frequently Bought Together