Order Entry
Denmark
ContactUsLinkComponent
Mouse Recombinant IL-13
Mouse Recombinant IL-13
Certificates
undefined
Mouse Recombinant IL-13

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Protein/peptide name:
    IL-13
  • Purity:
    ≥95%

Specifications

Frequently Bought Together