Order Entry
Denmark
ContactUsLinkComponent
Human Recombinant IL-8 (CXCL8)
Human Recombinant IL-8 (CXCL8)
Catalog # STMC78084.1
CAS Number:  
undefined
Human Recombinant IL-8 (CXCL8)
Catalog # STMC78084.1
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    IL-8 (CXCL8)
  • Purity:
    ≥95 %

Specifications

About this item

Interleukin-8 (IL-8) is a member of the CXC subfamily of chemokines and is produced by leukocytic cells (monocytes, T cells, neutrophils, and natural killer cells) and non-leukocytic somatic cells (endothelial cells, fibroblasts, and epithelial cells), with the most prominent source being monocytes and macrophages. Its production is induced by inflammatory stimuli, such as IL-1. IL-8, also known as CXCL8, activates neutrophils inducing chemotaxis, exocytosis, and the respiratory burst (Baggiolini and Clark-Lewis; Mukaida). IL-8 is considered one of the most potent neutrophil chemoattractants in inflammation and binds to two different chemokine receptors on leukocytes: the G protein-coupled receptors CXCR1 and CXCR2 (Hoffmann et al.; de Oliveira et al.). IL-8 has angiogenic effects on human intestinal microvascular endothelial cells in vitro that are mediated by CXCR2 (Heidemann et al.). IL-8 is reported to promote breast cancer progression by increasing cell invasion, angiogenesis, and metastasis and has been reported to be involved in regulating breast cancer stem-like cells (Singh et al.). IL-8 also has proangiogenic properties in inflammatory diseases of the conjunctiva, cornea, iris, retina, and orbit (Ghasemi et al.). It was also shown that a major T cell effector function in human newborns is IL-8 production, which has the potential to activate antimicrobial neutrophils and gamma/delta T cells (Gibbons et al.). A variety of human pathogens, such as HIV and Mycobacterium tuberculosis, have been shown to induce IL-8 production by monocytes and macrophages (Friedland et al.; Meddows-Taylor et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').