Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Human Recombinant Complement Factor D, His Tag

50 µG
  STMC100-1330
Marketsource
Human Recombinant Complement Factor D, His Tag
  STMC100-1330
Marketsource
 :  
Human Recombinant Complement Factor D, His Tag
Human Recombinant Complement Factor D, His Tag

50 µG

 :  
 
4.804,00 kr
   
1 * 1 Each
 

Complement factor D is a component of the alternative pathway of the complement system and part of the innate immune system, playing a vital role in the initiation and amplification of complement activation, in order to defend against infection (Barratt and Weitz). A serine protease belonging to the S1 peptidase family, complement factor D is secreted by adipocytes into circulating blood, and is also expressed by macrophages and monocytes (White et al.). In the initiation phase of the complement pathway, complement factor D cleaves complement factor B (bound to component C3) to produce a complex known as C3 convertase. During the amplification phase, complement factor D cleaves complement factor B (bound to component C3b) to produce the C3bBb convertase, and is involved in the propagation of complement activation. In addition to its immunological role, complement factor D is involved in other physiological processes, such as the efficient clearing of damaged cell debris by phagocytes following acute liver injury (Cresci et al.). Complement factor D deficiency is associated with an increased susceptibility to pathogens like Neisseria meningitidis (Biesma et al.). This protein contains a His-residue tag at the carboxyl end of the polypeptide chain. For consistency and reproducibility across your applications, complement factor D from STEMCELL comes lyophilised with ≥94% purity, and is verified by LAL analysis to ensure endotoxin levels are ≤1,0 EU/μg protein.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

 

 

  • Pk:
    50 µG
  • Cat. No.:
    STMC100-1330
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    Complement Factor D
  • Protein synonyms:
    Adipsin|ADN|DF|Factor D protein|PFD
  • Purity:
    ≥94%
  • Species:
    Human

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.