Order Entry
Denmark
ContactUsLinkComponent
Human Recombinant FGF-10 (KGF-2)
Human Recombinant FGF-10 (KGF-2)
Certificates
undefined
Human Recombinant FGF-10 (KGF-2)

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    FGF-10 (KGF-2)
  • Purity:
    ≥95%

Specifications

Frequently Bought Together