Order Entry
Denmark
ContactUsLinkComponent
Human/Mouse Recombinant Activin A
Human/Mouse Recombinant Activin A
Certificates
undefined
Human/Mouse Recombinant Activin A

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
    Mouse
  • Protein/peptide name:
    Activin A
  • Purity:
    ≥95%

Specifications

Frequently Bought Together