Order Entry
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant CNTF

Human Recombinant CNTF

Marketsource
Human Recombinant CNTF
Marketsource
Certificates
Human Recombinant CNTF

2 Options Available Below

Product Details & Documents

Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that belongs to the four-helix bundle cytokine family and is structurally related to interleukin 6 (IL-6), interleukin 11 (IL-11), leukemia inhibitory factor (LIF), and oncostatin M (OSM). CNTF binds to its receptor CNFTRα and induces formation of a heterodimer of the signal-transducing IL-6 receptor gp130 and LIF receptor (LIFR)-β, which triggers JAK/STAT, ERK, and the PI3K signaling cascades (Schuster et al.). CNTF plays an important role in neurogenesis and the differentiation of neural stem cells and has been suggested to possess a therapeutic role in treating neurological disorders (Ding et al.; Oppenheim et al.). CNTF has also been shown to protect rod photoreceptors from light-induced damage and to have therapeutic effects on retinal degenerative diseases caused by genetic defect or damage induced by toxins, autoantibodies, or strong light (Pernet et al.; Rhee et al.). Another therapeutic role of CNTF has been reported in protecting oligodendrocytes from death induced by apoptosis (Louis et al.). Additionally, CNTF is commonly used to differentiate human pluripotent stem cell (hPSC)-derived neural progenitor cells into astrocytes (Krencik and Zhang).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • STMC78010
  • STMC78010.1
  • Protein/peptide name:
  • CNTF
  • CNTF
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Purity:
  • ≥95%
  • ≥95%

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 10 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 1.339,00 kr
    1.339,00 kr
    1 * 1 Each
       1.339,00 kr

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC78010
    Protein/peptide name
    CNTF
    Protein/peptide type
    Recombinant
    Species
    Human
    Purity
    ≥95%
    Marketsource
  • 50 µG
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 3.025,00 kr
    3.025,00 kr
    1 * 1 Each
       3.025,00 kr

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC78010.1
    Protein/peptide name
    CNTF
    Protein/peptide type
    Recombinant
    Species
    Human
    Purity
    ≥95%
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.