Order Entry
Denmark
Orders LinkContactUsLinkComponent
EEB Expansion

EEB Expansion

EEB expansion, 6, 6 Assays
Catalog # OXNTEXP-EEB001
EEB Expansion
Catalog # OXNTEXP-EEB001
CAS Number:  
EEB Expansion
EEB Expansion

EEB expansion, 6, 6 Assays

CAS Number:  
Price
255,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

Additional Extraction EB (EEB) for use with the Ultra-long DNA sequencing kit V14 (SQK-ULK114).

Delivery information: This product is stable during shipping for seven days at temperatures up to 25 °C. Upon receipt, please store the expansion under the recommended conditions specified on the expansion label. Oxford Nanopore Technologies deem the useful life of the product to be three months from receipt by the customer.

Caution: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.

Specifications

Specifications

  • Pk:
    6 Assays
  • Description:
    EEB expansion
  • No. of reactions:
    6

Specifications