Order Entry
Denmark
ContactUsLinkComponent
 
Coomassie® brilliant blue R-250 in methanol for SDS-PAGE, Sigma-Aldrich®
Catalog # B8647-1EA
CAS Number:  
undefined
Coomassie® brilliant blue R-250 in methanol for SDS-PAGE, Sigma-Aldrich®
Catalog # B8647-1EA
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Appearance (Colour):
    Dark Blue
  • Appearance (Form):
    Liquid
  • Suitability, Suitable for use as a protein stain in electrophoresis:
    Pass

Specification Test Results

About this item

 :
 :825.98 g/mol
ADR:3 (6.1),II
 :M-Clarity™ quality level = MQ200
Flash Pt:14 °C (closed cup)
UN:1230
 :GHS02
 :GHS06
 :GHS08
Storage Temperature:Room Temperature
CAS Number:6104-59-2
Formula:C₄₅H₄₄N₃NaO₇S₂
MDL Number:MFCD00041762