Order Entry
Denmark
ContactUsLinkComponent
Fluorescein isothiocyanate (mixture of 5 and 6 isomers) ≥90% (by HPLC), Sigma-Aldrich®
Fluorescein isothiocyanate (mixture of 5 and 6 isomers) ≥90% (by HPLC), Sigma-Aldrich®
Catalog # F3651-100MG
CAS Number:  
Fluorescein isothiocyanate (mixture of 5 and 6 isomers) ≥90% (by HPLC), Sigma-Aldrich®
Catalog # F3651-100MG
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance (Colour):
    Yellow to Orange
  • Appearance (Form):
    Powder
  • Solubility (Colour):
    Very Faint Yellow to Yellow and Very Faint Green-Yellow to Green-Yellow
  • Solubility (Turbidity), 1 mg/ml, Acetone:
    Clear
  • 1H NMR Spectrum:
    Conforms to Structure
  • Water (by Karl Fischer):
    ≤6.5%
  • Purity (HPLC), Based on Isomer I and Isomer II:
    ≥90%
  • Carbon (anhydrous):
    54.1 - 71.0%
  • Nitrogen (anhydrous):
    3.0 - 4.0%

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ200
 :389.38 g/mol
Storage Temperature:Refrigerator
EINECS:248-207-7
CAS Number:27072-45-3
Formula:C₂₁H₁₁NO₅S
MDL Number:MFCD00005063