Order Entry
Denmark
Orders LinkContactUsLinkComponent
Crystal violet

Crystal violet

Corrosive
Health
Irritant
Environment
Supplier:   Merck
Crystal violet
Supplier:  Merck
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
  • CAS Number:548-62-9
  • Formula:C₂₅H₃₀ClN₃
  • MDL Number:MFCD00011750
  • Molecular Weight:407.99 g/mol
  • Merck Index:13,04406
  • Melting Pt:189…194 °C
  • Density:1.19 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:208-953-6
  • UN:3077
  • ADR:9,III
  •  :M-Clarity™ quality level = MQ200

Documents

Specifications for Product Family

Specification Test Results

  • Solubility:
    10 g/l (20 °C)
  • Melting Point:
    189 - 194 °C
  • Molar Mass:
    407.99 g/mol
  • Bulk Density:
    220 - 400 kg/m3
  • Density:
    1.19 g/cm3 (20 °C)
  • pH:
    2.5 - 3.5 (10 g/l, H2O, 20 °C)
  • Hazard Class:
    9
  • Hazard Class Description:
    Miscellaneous Hazardous Material
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.