



Crystal violet, Sigma-Aldrich®
Leverandør: SIGMA ALDRICH MICROSCOPY
Certifikater
Om denne vare
CAS nummer:548-62-9
:
Formel:C₂₅H₃₀ClN₃
MDL nummer:MFCD00011750
:407.99 g/mol
Merck Index:13,04406
Smeltende Pt:189…194 °C
Tæthed:1.19 g/cm³ (20 °C)
EINECS:208-953-6
FN:3077
ADR:9,III
:GHS05
:GHS08
:GHS07
:GHS09
Opbevaringstemperatur:Ambient
Relateret information
Frequently Bought Together