
Eosin Y (yellowish), pure, certified
Catalog # ACRO409430250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Red to brown Powder
- Infrared spectrum:Conforms
- Dye content:≥88 %
- UV Lambda max:515 to 518 nm
- Absorption Ratio P-15/P+15:1.21 to 1.77
- BSC Certification:Passes
Specification Test Results
About this item
:
Merck Index:13,03634
Melting Pt:>300 °C (dec.)
:691.86 g/mol
EINECS:241-409-6
:GHS07
Storage Temperature:Ambient
CAS Number:17372-87-1
Formula:C₂₀H₆Br₄Na₂O₅
MDL Number:MFCD00005040