Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Eosin Y (yellowish)
Eosin Y (yellowish)
Eosin Y (yellowish)
:
Thermo Fisher Scientific
Eosin Y (yellowish)
:
Thermo Fisher Scientific
:
17372-87-1
:
:
C₂₀H₆Br₄Na₂O₅
:
MFCD00005040
:
14,3603
:
295…296 °C
:
691.86 g/mol
:
241-409-6
:
GHS07
:
Ambient
msds_ALFAB24535
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Test strips for peracetic acid, MQuant®, Supelco®
: Merck
pH indicator papers, MQuant®, Supelco®
: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
: VWR Chemicals
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Test kits, nitrite, Spectroquant®, Supelco®
: Merck
VWR® Rota®, pH indicator paper, 1 to 14
: VWR Chemicals
Mercury standard solution, 1000 mg/L Hg in 2 M nitric acid (from Hg(NO₃)₂) Certipur® standard for AAS, Supelco®
: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
: Merck
Test kits, phosphate, Spectroquant®, Supelco®
: Merck
Test strips for peroxide, MQuant®, Supelco®
: Merck
Sodium hypochlorite (3,5% Cl₂) in aqueous solution, GPR RECTAPUR®
: VWR Chemicals
Test kits, ammonium, Spectroquant®, Supelco®
: Merck
pH indicator strips, MQuant®, Supelco®
: Merck
Citric acid monohydrate 99.0-102.0% ACS
: Thermo Fisher Scientific