Order Entry
Denmark
Orders LinkContactUsLinkComponent
Fuchsin (acid)

Fuchsin (acid)

Glass bottle for solids, 10 g
Catalog # ROTH4268.5
Marketsource
Supplier:   Roth Carl
Fuchsin (acid)
Catalog # ROTH4268.5
Supplier:  Roth Carl
Marketsource
CAS Number:  3244-88-0
Fuchsin (acid)

Glass bottle for solids, 10 g

CAS Number:  3244-88-0
Price
Inquire For Price
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Rubin S, CN DcR-8, Acid Violet 19, Acid Fuchsin, Acid Fuchsin sodium salt, Fuchsin S, CI 42685, Acid Magenta, 3-(1-(4-Amino-3-methyl-5-sulphonatophenyl)-1-(4-amino-3-sulphonatophenyl)methylene)cyclohexa-1,4-dienesulphonic acid, Benzenesulphonic acid, 2-amino-5-[(4-amino-3-sulphophenyl)(4-imino-3-sulpho-2,5-cyclohexadien-1-ylidene)methyl]-3-methyl-, sodium salt (1:2)
  • CAS Number:3244-88-0
  • Formula:C₂₀H₁₇N₃Na₂O₉S₃
  • MDL Number:MFCD00013286
  • Molecular Weight:585.55 g/mol
  • Merck Index:12,00109
  • Melting Pt:> 130 °C
  • Storage Temperature:Ambient
  • EINECS:221-816-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.