Fuchsin (acid)
Supplier: Thermo Fisher Scientific
Jump to:
About this item
CAS No.: 3244-88-0 Molecular Formula: C20H17N3Na2O9S3 Formula Weight: 585.53 Beilstein Reference: 4111656 Merck Reference: 14,107 MDL No.: MFCD00013286
Synonyms: Rubin S, CN DcR-8, Acid Violet 19, Acid Fuchsin, Acid Fuchsin sodium salt, Fuchsin S, CI 42685, Acid Magenta, 3-(1-(4-Amino-3-methyl-5-sulphonatophenyl)-1-(4-amino-3-sulphonatophenyl)methylene)cyclohexa-1,4-dienesulphonic acid, Benzenesulphonic acid, 2-amino-5-[(4-amino-3-sulphophenyl)(4-imino-3-sulpho-2,5-cyclohexadien-1-ylidene)methyl]-3-methyl-, sodium salt (1:2)
- CAS Number:3244-88-0
- Formula:C₂₀H₁₇N₃Na₂O₉S₃
- MDL Number:MFCD00013286
- Molecular Weight:585.55 g/mol
- Merck Index:14,107
- Melting Pt:> 130 °C
- Storage Temperature:Ambient
- EINECS:221-816-5
3 Options Available Below
Product Details & Documents
CAS No.: 3244-88-0 Molecular Formula: C20H17N3Na2O9S3 Formula Weight: 585.53 Beilstein Reference: 4111656 Merck Reference: 14,107 MDL No.: MFCD00013286
Synonyms: Rubin S, CN DcR-8, Acid Violet 19, Acid Fuchsin, Acid Fuchsin sodium salt, Fuchsin S, CI 42685, Acid Magenta, 3-(1-(4-Amino-3-methyl-5-sulphonatophenyl)-1-(4-amino-3-sulphonatophenyl)methylene)cyclohexa-1,4-dienesulphonic acid, Benzenesulphonic acid, 2-amino-5-[(4-amino-3-sulphophenyl)(4-imino-3-sulpho-2,5-cyclohexadien-1-ylidene)methyl]-3-methyl-, sodium salt (1:2)
- CAS Number:3244-88-0
- Formula:C₂₀H₁₇N₃Na₂O₉S₃
- MDL Number:MFCD00013286
- Molecular Weight:585.55 g/mol
- Merck Index:14,107
- Melting Pt:> 130 °C
- Storage Temperature:Ambient
- EINECS:221-816-5



