Thiamine hydrochloride 98.5-101.5%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride
CAS Number:67-03-8
:
Formula:C₁₂H₁₈Cl₂N₄OS
MDL Number:MFCD00012780
Molecular Weight:337.27 g/mol
Melting Pt:250 °C (dec.)
Merck Index:13,09366
EINECS:200-641-8
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Water:≤5 % (K.F.)
- Sulfated ash:≤0.1 %
- Titration with HClO4:98.5 to 101.5 % (On anhydrous substance)
- Sulfate (SO4):≤300 ppm
- Heavy metals (as Pb):≤20 ppm
Specification Test Results
Frequently Bought Together