Order Entry
Denmark
ContactUsLinkComponent
1,1-Dimethoxy-3-nitropropane
1,1-Dimethoxy-3-nitropropane
Catalog # 8.14137.0001
Supplier:  Merck
CAS Number:  
1,1-Dimethoxy-3-nitropropane
Catalog # 8.14137.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 ml
  • Seq:
    2070

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Molar Mass:
    149.14 g/mol
  • Boiling Point:
    96 °C (20 hPa)
  • Flash Point:
    90 °C
  • Density:
    1.12 g/cm3 (20 °C)
  • Synonyms:
    3-Nitropropionaldehyde dimethyl acetal
  • Hill Formula:
    C5H11NO4
  • Chemical Formula:
    O2N-CH2-CH2-CH(OCH3)2

Specification Test Results

About this item

 :
 :149.15 g/mol
Density:1.12 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
CAS Number:72447-81-5
Formula:C₅H₁₁NO₄