Calcium Standard Solution, 1000 mg/L as Ca (NIST), 500 ml, Hach
Catalog # HACH2240349
Supplier: Hach
New Product
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O
- Pk:500 mL
- Certification:NIST
- Element:Calcium
Specifications
About this item
For accuracy checks and instrument calibrations in calcium determinations.