Order Entry
Denmark
ContactUsLinkComponent
Eosin Y solution, alcoholic , with Phloxin
Flammable
Irritant
Health
Eosin Y solution, alcoholic , with Phloxin
Catalog # HT1103128-4L
CAS Number:  
Flammable
Irritant
Health
Eosin Y solution, alcoholic , with Phloxin
Catalog # HT1103128-4L
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Appearance (Colour)
    Bright pink to pinkish-orange:
    Conforms
  • Appearance (Form):
    Solution
  • Appearance (Turbidity):
    Clear
  • Performance:
    Conforms
  • Expiration Date Period:
    24 months

Specification Test Results

About this item

 :
 :647.89 g/mol
 :M-Clarity™ quality level = MQ500 (more info.)
Flash Pt:13 °C (closed cup)
 :GHS02
 :GHS07
 :GHS08
Storage Temperature:Room Temperature
CAS Number:15086-94-9
Formula:C₂₀H₈Br₄O₅
MDL Number:MFCD00036189