Eosin Y solution Aqueous
4 L
Catalog # HT1102128-4L
Supplier: SIGMA ALDRICH MICROSCOPY
Eosin Y solution Aqueous
Catalog # HT1102128-4L
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number: 15086-94-9
Jump to:
Product Details & Documents
General purpose cytoplasmic counterstain. Used with hematoxylin and eosin staining.
Certifications: Certified Eosin Y, 0.5% (w/v) in water. Not acidified.
Synonyms: Acid Red 87, Tetrabromofluorescein, 2-(3,6-Dihydroxy-2,4,5,7-tetrabromoxanthen-9-yl)-benzoic acid, 2,4,5,7-Tetrabromofluorescein, Eosin yellowish, Solvent red 43, Bromofluorescein, Bromo acid J. TS, XL, or XX, CI 45380, Bronze Bromo ES, Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 2',4',5',7'-tetrabromo-3',6'-dihydroxy-
- CAS Number:15086-94-9
- Formula:C₂₀H₈Br₄O₅
- MDL Number:MFCD00036189
- Molecular Weight:647.89 g/mol
- :M-Clarity™ quality level = MQ500 (more info.)
- Storage Temperature:Room Temperature
Documents
Merck The Color of Tissue Diagnostics
Product Literature
Specifications
Specification Test Results
- Appearance (Colour)
Greenish- Pink-Orange:Conforms - Appearance (Form):Solution
- Appearance (Turbidity)
Essentially free of precipitate:Clear - Performance:Conforms
- Expiration Date Period:18 months
Specifications
- Pk:4 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



