Nile red, Technical Grade, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:7385-67-3
:
Formula:C₂₀H₁₈N₂O₂
MDL Number:MFCD00011639
:318.38 g/mol
Melting Pt:203…205 °C (lit.)
:M-Clarity™ quality level = MQ200
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Colour):Faint Green to Brown
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Carbon:70.0 - 77.3 %
- Nitrogen:7.5 - 8.8 %
- Solubility (Colour):Faint Pink to Very Dark Red
- Solubility (Turbidity)
1 mg/mL, MeOH:Clear to Turbid
Specification Test Results
Frequently Bought Together