Eosin Y solution Aqueous, Sigma-Aldrich®
1 L
Katalog # HT110232-1L
Leverandør: SIGMA ALDRICH MICROSCOPY
Eosin Y solution Aqueous, Sigma-Aldrich®
Katalog # HT110232-1L
Leverandør: SIGMA ALDRICH MICROSCOPY
CAS nummer: 15086-94-9
Gå til:
Product Details & Documents
General purpose cytoplasmic counterstain. Used with hematoxylin and eosin staining.
Certifications: Certified Eosin Y, 0.5% (w/v) in water. Not acidified.
Synonymer: Acid Red 87, Tetrabromofluorescein, 2-(3,6-Dihydroxy-2,4,5,7-tetrabromoxanthen-9-yl)-benzoic acid, 2,4,5,7-Tetrabromofluorescein, Eosin yellowish, Solvent red 43, Bromofluorescein, Bromo acid J. TS, XL, or XX, CI 45380, Bronze Bromo ES, Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 2',4',5',7'-tetrabromo-3',6'-dihydroxy-
- CAS nummer:15086-94-9
- Formel:C₂₀H₈Br₄O₅
- MDL nummer:MFCD00036189
- Molecular Weight:647.89 g/mol
- :M-Clarity™ quality level = MQ500 (more info.)
- Opbevaringstemperatur:Room Temperature
Documents
Merck The Color of Tissue Diagnostics
Product Literature
Specifikationer
Specifikationstestresultater
- Appearance (Colour)
Greenish- Pink-Orange:Conforms - Appearance (Form):Solution
- Appearance (Turbidity):Clear
- Performance:Conforms
- Expiration Date Period:18 months
Specifikationer
- Pk:1 L
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



