Order Entry
Denmark
ContactUsLinkComponent
Safranine O solution 0.6%, Sigma-Aldrich®
Safranine O solution 0.6%, Sigma-Aldrich®
Catalog # HT90432-1L
CAS Number:  
Safranine O solution 0.6%, Sigma-Aldrich®
Catalog # HT90432-1L
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Appearance (Colour):
    Red
  • Appearance (Form):
    Solution
  • Essentially free of particles/precipitate:
    Clear
  • Performance:
    Conforms
  • Expiration Date Period:
    24 months

Specification Test Results

About this item

 :
 :350.85 g/mol
 :M-Clarity™ quality level = MQ500 (more info.)
Flash Pt:37.8 °C (closed cup)
 :GHS02
Storage Temperature:Room Temperature
CAS Number:477-73-6
Formula:C₂₀H₁₉ClN₄
MDL Number:MFCD00011759