Order Entry
Denmark
Orders LinkContactUsLinkComponent
Hydroquinone

Hydroquinone

10 Tab.
Catalog # HT1014-10TAB
Corrosive
Irritant
Health
Environment
Hydroquinone
Catalog # HT1014-10TAB
CAS Number:  123-31-9
Hydroquinone
Hydroquinone

10 Tab.

CAS Number:  123-31-9
Price
1.130,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

Intended for use in the Silver Stain (Steiner-Steiner) kit. Hydroquinone, 0.5 g each, with excipients.

Synonyms:  1,4-Dihydroxybenzene, Quinol
  • CAS Number:123-31-9
  • Formula:C₆H₄(OH)₂
  • MDL Number:MFCD00002339
  • Molecular Weight:110.11 g/mol
  •  :M-Clarity™ quality level = MQ500 (more info.)
  • Flash Pt:165 °C (closed cup)
  • Storage Temperature:Refrigerator
Specifications

Specification Test Results

  • Appearance (Colour):
    White to Off White
  • Appearance (Form)
    Intact:
    Round Tablet
  • Performance:
    Conforms
  • Expiration Date Period:
    24 months

Specifications

  • Pk:
    10 Tab.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.