Gold chloride solution 200 mg/dl in deionised water
HT1004-100ML
:
- Pk:100 mL
- Seq:2440
- Appearance (Colour):Yellow
- Appearance (Form):Solution
- Appearance (Turbidity)
Essentially free of particles/precipitate:Clear - Performance:Conforms
- undefined:When using with Sigma-Aldrich Procedure No. HT100, fungi stain purplish brown to black.