Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Formaldehyde solution 4%

Health
Irritant
Leverandør:   Ampliqon A/S
Formaldehyde solution 4%
Leverandør:  Ampliqon A/S
Certificates
Formaldehyde solution 4%
Formaldehyde solution 4%

3 Options Available Below

Product Details & Documents

(10% Formaline solution)

Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-001-8
Product Family Options

Product Information