ESD Safety Ankle Boots, Lacing Sytem, Mike Boa® Mid S3 S
: ELTEN
ESD safety ankle boot made from hydrophobised microfibre and textile upper and a PU/PU sole (Crossworker).
- Fabric lining for natural ventilation
- Full, padded bellows tongue
- Full-length inlay sole ESD PRO black
- Boa® fit lacing system
- Non-metallic, penetration-resistant midsole
- Steel toe cap
- PU scuff cap
Suitable for areas where there is a risk of electrostatic discharge (ESDS/ESD).
Colour: Black
: EN ISO 20345 S3S FO/SR/SC/LG, form B.
- Catalog No:
- ELTN763831/42
- ELTN763831/43
- ELTN763831/45
- ELTN763831/46
- ELTN763831/44
- ELTN763831/47
- ELTN763831/48
- ELTN763831/40
- ELTN763831/41
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
Frequently Bought Together