Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (iFluor® 594)
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
: For research use only.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: L17F12
Conjugation: iFluor® 594
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
- Catalog No:
- AATB100510C0
- AATB100510C1
- Antigen symbol:
- CD5
- CD5
- Antigen name:
- CD5 antigen
- CD5 antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- iFluor® 594
- iFluor® 594
- Clone:
- L17F12
- L17F12
- Reactivity:
- Human
- Host:
- Mouse
- Mouse
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- Storage temperature:
- 2...8 °C with minimized light exposure. Do not freeze.
- 2...8 °C with minimized light exposure. Do not freeze.
- Shipping temperature:
- 20
- 20
Frequently Bought Together