Anti-ZC3H8 Rabbit Polyclonal Antibody
Catalog # PRSI55-863
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Zinc finger CCCH domain-containing protein 8
- Antigen symbol:ZC3H8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:Amino Acid Range = 52-81
- Form:Liquid
- Gene ID:Q8N5P1
- Antigen synonyms:ZC3H8
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:34 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,47 mg/ml
- Shipping temperature:4 °C
- Immunogen:This ZC3H8 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 52-81 amino acids from the N-terminal region of human ZC3H8.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Size:400 µl
- Pk:400 µl
Specifications
About this item
Acts as a transcriptional repressor of the GATA3 promoter. Sequence-specific DNA-binding factor that binds to the 5'-AGGTCTC-3' sequence within the negative cis-acting element intronic regulatory region (IRR) of the GATA3 gene (By similarity). Induces thymocyte apoptosis when overexpressed, which may indicate a role in regulation of thymocyte homeostasis.
For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: ZC3H8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse