Anti-PROC Rabbit Polyclonal Antibody
Catalog # PRSI15-483
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Protein C (inactivator of coagulation factors Va and VIIIa)
- Antigen symbol:PROC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:5624
- Antigen synonyms:APC|THPH4|PROC|PC|THPH3|PROC1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 182-461 of human PROC (NP_000303.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PROC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat