Order Entry
Denmark
ContactUsLinkComponent
Anti-AK8 Rabbit Polyclonal Antibody
Anti-AK8 Rabbit Polyclonal Antibody
Catalog # PRSI15-188
Supplier:  ProSci Inc.
Anti-AK8 Rabbit Polyclonal Antibody
Catalog # PRSI15-188
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Adenylate kinase 8
  • Antigen symbol:
    AK8
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    158067
  • Antigen synonyms:
    AK 8|adenylate kinase 8|C9orf98|ATP-AMP transphosphorylase 8|DDX31|putative adenylate kinase-like protein C9orf98
  • Modification:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at −20 °C. Avoid freeze/thaw cycles
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human AK8 (NP_689785.1)
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: AK8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat