Anti-AMER1 Rabbit Polyclonal Antibody
Katalog # PRSI62-320
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:APC membrane recruitment protein 1
- Antigen symbol:AMER1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:681-708 aa
- Form:Liquid
- Gene ID:139285
- Modification:None
- Molecular weight:124 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This WTX antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 681-708 amino acids from the central region of human WTX.
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis.
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: AMER1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 681-708
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse