Alcohol Plus Hand Sanitisers
Supplier: Rubbermaid Commercial Products
New Product
Certificates
About this item
Effective and fast-acting hand sanitiser is fully tested and proven to be bactericidal, virucidal, and yeasticidal and kills 99,99% of common germs.
- Fast-acting formulation disinfects hands in 30 seconds
- Contains 70% alcohol in line with WHO recommendations
- Contains moisturising ingredients to help prevent skin dryness
Certifications: The Alcohol Plus formulation has been tested against the following standards: EN 1500, EN 1276, EN 13727 + A2, EN 14476 + A1, EN 14476 + A2, EN 1650 + A1, and EN 13624.
Frequently Bought Together