Anti-POP7 Rabbit Polyclonal Antibody
Catalog # PRSI23-449
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:POP7 homolog, ribonuclease P/MRP subunit
- Antigen symbol:POP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:10248
- Antigen synonyms:0610037N12Rik|ribonuclease P/MRP subunit|ribonucleases P/MRP protein subunit POP7 homolog|RNaseP protein p2hPOP7|processing of precursor 7|RPP2ribonuclease P protein subunit p2POP7 (processing of precursor|S. cerevisiae) homolog|ribonuclease P subunit|RPP2
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human POP7 (NP_005828.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: POP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human