Anti-SLC35A2 Rabbit Polyclonal Antibody
PRSI22-801
New Product
- Antibody type:Primary
- Antigen name:Solute carrier family 35 member A2
- Antigen symbol:SLC35A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:7355
- Antigen synonyms:UGT2|solute carrier family 35 (UDP-galactose transporter)|UDP-galactose translocator|CDGX|CDG2M|member A2|member 2|UGTL|UGT|UGAT|UDP-Gal-Tr|UGT1|UGALT
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-242 of human SLC35A2 (NP_001027460.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: SLC35A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat