Order Entry
Denmark
ContactUsLinkComponent
Anti-S Rabbit Recombinant Antibody [clone: C-A11]
Anti-S Rabbit Recombinant Antibody [clone: C-A11]
Catalog # PRSI20-186
Supplier:  ProSci Inc.
Anti-S Rabbit Recombinant Antibody [clone: C-A11]
Catalog # PRSI20-186
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Surface glycoprotein
  • Antigen symbol:
    S
  • Clonality:
    Recombinant
  • Clone:
    C-A11
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Virus
  • Neutralisation:
    Yes
  • Form:
    Liquid
  • Gene ID:
    43740568
  • Antigen synonyms:
    South Africa variant B.1.351|U.K variant B.1.1.7|Binding to ACE-2 receptor|SARS-CoV-2 variants|Spike protein S1|RBD domain|Spike protein S2
  • Storage buffer:
    1X PBS
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    recombinant protein (RBD).
  • Purification:
    Protein A
  • Pack type:
    Vial
  • Pk:
    200 µl

Specifications

About this item

Storage temperature: SARS-CoV-2 (COVID-19) Spike RBD neutralizing antibody [C-A11] can be stored at 4...730 °C for three months and −20...+730 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.

Type: Primary
Antigen: S
Clonality: Recombinant Monoclonal
Clone: C-A11
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1 kappa
Reactivity: Virus