Order Entry
Denmark
ContactUsLinkComponent
 
Anti-NAAA Rabbit Polyclonal Antibody
  PRSI59-962
New Product
undefined
Anti-NAAA Rabbit Polyclonal Antibody
  PRSI59-962
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    N-acylethanolamine-hydrolyzing acid amidase
  • Antigen symbol:
    NAAA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    331 - 359 aa
  • Form:
    Liquid
  • Gene ID:
    27163
  • Modification:
    None
  • Molecular weight:
    40 kDa
  • Storage temperature:
    Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4°C
  • Immunogen:
    This NAAA antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 331 - 359 amino acids from the C-terminal region of human NAAA.
  • Purification:
    This antibody is purified through a protein A column, followed by peptide affinity purification
  • Pk:
    400 µl

 

 

For WB starting dilution is: 1:1000.

Type: Primary
Antigen: NAAA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 331-359
Host: Rabbit
Isotype: Ig
Reactivity: Human