Order Entry
Denmark
ContactUsLinkComponent
 
Human Recombinant RELT (from Cells)
Catalog # PRSI91-807
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant RELT (from Cells)
Catalog # PRSI91-807
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    0,05 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimise freeze-thaw cycles.
  • Endotoxin content:
    <0,1 ng/µg (1 IEU/µg) as determined by LAL test
  • Protein synonyms:
    Tumor necrosis factor receptor superfamily member 19L|TNFRSF19L|Receptor expressed in lymphoid tissues|RELT
  • Protein/peptide name:
    RELT
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    41,4 kDa
  • Sequence:
    Ser26-Ala160
  • Formulation:
    Supplied as a 0,2 µm filtered solution of 20 mM PB, 150 mM NaCl, pH 7,2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 µg/ml. Dissolve the Lyophilised protein in ddH₂O. Please aliquot the reconstituted solution to minimise freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Tumor necrosis factor receptor superfamily member 19L (TNFRSF19L), also known as Receptor expressed in lymphoid tissues and RELT, is a member of the TNF-receptor superfamily. TNFRSF19L is a single-pass type membrane protein and contains one TNFR-Cys repeat. TNFRSF19L is highly expressed in spleen, lymph node, thymus, peripheral blood leukocytes, bone marrow and fetal liver. It has been shown TNFRSF19L activates the NF-kappaB pathway and selectively binds TNF receptor-associated factor 1 (TRAF1). TNFRSF19L is capable of stimulating T-cell proliferation in the presence of CD3 signaling, which suggests its regulatory role in immune response.

  • Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays.

Caution: For research use only.