Order Entry
Denmark
ContactUsLinkComponent
Anti-CR2 Mouse Monoclonal Antibody [clone: HB5] (PE (Phycoerythrin))
Anti-CR2 Mouse Monoclonal Antibody [clone: HB5] (PE (Phycoerythrin))
Catalog # PRSI76-156
Supplier:  ProSci Inc.
New Product
Anti-CR2 Mouse Monoclonal Antibody [clone: HB5] (PE (Phycoerythrin))
Catalog # PRSI76-156
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    complement component (3d/Epstein Barr virus) receptor 2
  • Antigen symbol:
    CR2
  • Clonality:
    Monoclonal
  • Clone:
    HB5
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    P20023
  • Antigen synonyms:
    CR2
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    100 Tests

Specifications

About this item

The HB5 monoclonal antibody specifically reacts with human CD21, a 145 kDa glycosylated type I integral membrane protein. CD21 is reported to be involved in B cell activation and proliferation. It is expressed on follicular dendritic cells, mature B cells, a subset of T cells, and a subset of epithelial cells. It is the receptor utilized by the Epstein-Barr virus to infect B cells.

Type: Primary
Antigen: CR2
Clonality: Monoclonal
Clone: HB5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human