Anti-Lgals9 Recombinant Antibody [Clone: RG9-35]
PRSI24-626
New Product
- Antibody type:Primary
- Antigen name:lectin, galactose binding, soluble 9
- Antigen symbol:Lgals9
- Clonality:Recombinant
- Clone:RG9-35
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:16859
- Antigen synonyms:ecalectin|tumor antigen HOM-HD-21|soluble 5|HUAT|Galectin-9|urate transporter/channel protein.|LGALS9A|galactose binding|galectin 9|Gal-9|lectin
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising rats with recombinant mouse galectin-9
- Tested applications:Blocking, functional assays
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: lectin, galactose binding, soluble 9
Clonality: Recombinant
Clone: RG9-35
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a kappa
Reactivity: Mouse